Learn More
ABCG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB400-117
This item is not returnable.
View return policy
Description
ABCG8 Polyclonal specifically detects ABCG8 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Immunocytochemistry, Immunofluorescence | |
| Western Blot 1:250-1:500, Immunocytochemistry/Immunofluorescence 1:250 | |
| ABCG8 | |
| IgG |
| Polyclonal | |
| ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |