Learn More
ABCG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand:  Novus Biologicals NB400-117
 View more versions of this product
Product Code. 18711684
  • 493.00€ / 0.20mL
Description
ABCG8 Polyclonal specifically detects ABCG8 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry, Immunofluorescence
Western Blot 1:250-1:500, Immunocytochemistry/Immunofluorescence 1:250
ABCG8
IgG
Polyclonal
ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.