missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB400-117SS
This item is not returnable.
View return policy
Description
ABCG8 Polyclonal specifically detects ABCG8 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ABCG8 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.02% Sodium Azide | |
| ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
| Rabbit | |
| 70 kDa | |
| 0.05 mL | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 64241 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:250-1:500, Immunocytochemistry/Immunofluorescence 1:250 | |
| Q9H221 | |
| ABCG8 | |
| A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProt# Q9H221] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction