accessibility menu, dialog, popup

UserName

CAS RN 97-56-3

CH3NNH2NCH3(E/Z)

CAS RN 97-56-3

IUPAC Name: 2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline
Synonyms: 4-o-tolylazo-o-toluidine
Molecular Weight (g/mol): 225.3
Molecular Formula: C14H15N3
InChi Key: PFRYFZZSECNQOL-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1N)N=NC1=CC=CC=C1C
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Color 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)