Learn More
Fast Garnet GBC base, Thermo Scientific Chemicals
Product Code. 15463319
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463319 100 g 100g
15473319 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15463319 Supplier Thermo Scientific Alfa Aesar Supplier No. J60574.22
Request Bulk or Custom Format
C.I. 11160
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C.I. 11160
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97-56-3
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.295
MDL Number MFCD00007733
InChI Key PFRYFZZSECNQOL-UHFFFAOYSA-N
Synonym C.I. 11160; Solvent Yellow 3
PubChem CID 7340
IUPAC Name 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
Specifications
Melting Point 101°C to 102°C
Linear Formula CH3C6H4N=NC6H3(CH3)NH2
Beilstein 6506005
Merck Index 14,420
Formula Weight 225.29
Odor Odorless
Color Brown/Orange
Quantity 100 g
Physical Form Powder
Chemical Name or Material Fast Garnet GBC base