Learn More
4-Amino-2',3-dimethylazobenzene, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 4-Amino-2,3-dimethylazobenzene |
| CAS | 97-56-3 |
| Analyte or Component Names | 4-Amino-2,3-dimethylazobenzene |
| Shipping Condition | +20°C |
| Molecular Formula | C14 H15 N3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzenamine, 2-methyl-4-[(2-methylphenyl)azo]- (9CI), C.I. Solvent Yellow 3 (8CI), o-Toluidine, 4-o-tolylazo- (6CI), 2-Methyl-4-[2-(2-methylphenyl)diazenyl]benzenamine, 2,3'-Dimethyl-4'-aminoazobenzene, 2-Aminoazotoluene, 2-Methyl-4-[(2-methylphenyl)diazenyl]amine, 2-Methyl-4-[(o-tolyl)azo]aniline, 2',3-Dimethyl-4-aminoazobenzene, 4-(o-Tolylazo)-o-toluidine, 4-Amino-2',3-dimethylazobenzene, Brasilazina Oil Yellow R, C.I. 11160, C.I. 11160B, Fast Garnet GBC Base, Fast Yellow AT, Fat Yellow B, Hidaco Oil Yellow, NSC 1797, NSC 26821, OAAT, Oil Yellow 21, Oil Yellow 2681, Oil Yellow 2R, Oil Yellow AT, Oil Yellow C, Oil Yellow I, Organol Yellow 2T, Solvent Yellow 3, Somalia Yellow R, o-AT, o-Aminoazotoluene |
| InChI Formula | InChI=1S/C14H15N3/c1-10-5-3-4-6-14(10)17-16-12-7-8-13(15)11(2)9-12/h3-9H,15H2,1-2H3/b17-16+ |
| SMILES | Cc1ccccc1N=Nc2ccc(N)c(C)c2 |
| IUPAC Name | 2-methyl-4-[(E)-(2-methylphenyl)diazenyl]aniline |
| Show More |