CAS RN 73231-34-2
CAS RN 73231-34-2
IUPAC Name:
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methanesulfonylphenyl)propan-2-yl]acetamide
Synonyms:
florfenicol
Molecular Weight (g/mol):
358.21
Molecular Formula:
C12H14Cl2FNO4S
InChi Key:
AYIRNRDRBQJXIF-NXEZZACHSA-N
SMILES:
CS(=O)(=O)C1=CC=C(C=C1)[C@@H](O)[C@@H](CF)NC(=O)C(Cl)Cl
1
–
2
of
2
results
1
Florfenicol, 98%, Thermo Scientific™
CAS: 73231-34-2 Molecular Formula: C12H14Cl2FNO4S Molecular Weight (g/mol): 358.205 MDL Number: MFCD00864834 InChI Key: AYIRNRDRBQJXIF-NXEZZACHSA-N PubChem CID: 114811 ChEBI: CHEBI:87185 IUPAC Name: 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide SMILES: CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O
2
Florfenicol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality