missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Florfenicol, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Florfenicol |
| CAS | 73231-34-2 |
| Analyte or Component Names | Florfenicol |
| Shipping Condition | +20°C |
| Molecular Formula | C12 H14 Cl2 F N O4 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Acetamide, 2,2-dichloro-N-[1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]-, [R-(R*,S*)]-, (-)-Florfenicol, 2,2-Dichloro-N-[(1S,2R)-1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]acetamide, Aquafen, Aquaflor, Flonicol, Florfenicol, Florgane, Floron, Nuflor, Sch 25298, Selectan, Topazone |
| InChI Formula | InChI=1S/C12H14Cl2FNO4S/c1-21(19,20)8-4-2-7(3-5-8)10(17)9(6-15)16-12(18)11(13)14/h2-5,9-11,17H,6H2,1H3,(H,16,18)/t9-,10-/m1/s1 |
| SMILES | CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@@H](CF)NC(=O)C(Cl)Cl |
| IUPAC Name | 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction