accessibility menu, dialog, popup

UserName

CAS RN 5234-68-4

IUPAC Name: 2-methyl-N-phenyl-5,6-dihydro-1,4-oxathiine-3-carboxamide
Synonyms: carboxin DMOC rtv vitavax dual murganic rpb vitavax 30-C kisvax vitaflo 250 cerevax vitavax Pro-Gro
Molecular Weight (g/mol): 235.3
Molecular Formula: C12H13NO2S
InChi Key: GYSSRZJIHXQEHQ-UHFFFAOYSA-N
SMILES: CC1=C(SCCO1)C(=O)NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (2)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)