Learn More
Method 525.2 Nitrogen/Phosphorus Pesticide Mix, 6-13 1000 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Carboxin∣Diazinon∣Disulfoton∣Fenamiphos∣Merphos∣Terbufos |
| CAS | 5234-68-4, 333-41-5, 298-04-4, 22224-92-6, 150-50-5, 13071-79-9 |
| Analyte or Component Names | Carboxin∣Diazinon∣Disulfoton∣Fenamiphos∣Merphos∣Terbufos |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Carboxin 1000 μg/mL∣Diazinon 1000 μg/mL∣Disulfoton 1000 μg/mL∣Fenamiphos 1000 μg/mL∣Merphos 1000 μg/mL∣Terbufos 1000 μg/mL |