Learn More
Carboxin, SPEX CertiPrep™
Product Code. 15216306 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15216306 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15216306 Supplier SPEX Certiprep Supplier No. S760
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Carboxin5234-68-41000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging 2mL amber prescored glass ampule
CAS 5234-68-4, 67-64-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C12H13NO2S
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key GYSSRZJIHXQEHQ-UHFFFAOYSA-N
SMILES CC1=C(SCCO1)C(=O)NC2=CC=CC=C2
IUPAC Name 6-methyl-N-phenyl-2,3-dihydro-1,4-oxathiine-5-carboxamide
Solvent or Matrix Acetone
Molecular Weight (g/mol) 235.301
PubChem CID 21307
Quantity 1 mL
ChEBI CHEBI:41412
Chemical Name or Material Carboxin
Show More Show Less
Product intended for laboratory use only