Learn More
Carboxine, TRC
Specifications
Specifications
| Chemical Name or Material | Carboxin |
| CAS | 5234-68-4 |
| Molecular Formula | C12 H13 N O2 S |
| Quantity | 1 g |
| Synonym | 1,4-Oxathiin-3-carboxanilide, 5,6-dihydro-2-methyl- (7CI,8CI), 5,6-Dihydro-2-methyl-N-phenyl-1,4-oxathiin-3-carboxamide, 1-Oxa-2-methyl-3-(anilidocarbonyl)-4-thiacyclohex-2-ene, 2,3-Dihydro-5-carboxanilido-6-methyl-1,4-oxathiin, 2,3-Dihydro-6-methyl-1,4-oxathiin-5-carboxanilide, 2,3-Dihydro-6-methyl-1,4-oxathiine-5-carboxylic acid anilide, 2,3-Dihydro-6-methyloxathiin-5-carboxanilide, 5,6-Dihydro-2-methyl-1,4-oxathiin-3-carboxanilide, Carbathiin, Carboxin, Carboxine, D 735, DCMO, DMOC, F 735, Fenoxan, Karboxyn, N-Phenyl-2-methyl-5,6-dihydrooxathiin-3-carboxamide, NSC 263492, V 4X, Vitaflo 250, Vitavax, Vitavax 100, Vitavax 34, Vitavax 735D, Vitavax 75W, Vitavax EC 200 |
| InChI Formula | InChI=1S/C12H13NO2S/c1-9-11(16-8-7-15-9)12(14)13-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3,(H,13,14) |
| SMILES | CC1=C(SCCO1)C(=O)Nc2ccccc2 |
| IUPAC Name | 6-methyl-N-phenyl-2,3-dihydro-1,4-oxathiine-5-carboxamide |
| Molecular Weight (g/mol) | 235.3 |
| Formula Weight | 235.0667 |