accessibility menu, dialog, popup

UserName

CAS RN 29678-81-7

OH(R)OHO

CAS RN 29678-81-7

IUPAC Name: (2R)-2-hydroxy-4-phenylbutanoic acid
Synonyms: (2R)-2-hydroxy-4-phenylbutanoic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: JNJCEALGCZSIGB-SECBINFHSA-N
SMILES: O[C@H](CCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results