missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
| Antigen | ZSCAN2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ZSCAN2 Polyclonal antibody specifically detects ZSCAN2 in Human samples. It is validated for Western BlotSpecifications
| ZSCAN2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ20595, zfp-29, ZFP29, zinc finger and SCAN domain containing 2, zinc finger and SCAN domain-containing protein 2, Zinc finger protein 29 homolog, Zinc finger protein 854, ZNF854zinc finger protein 29 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human ZSCAN2 (NP_870992.2). GPESEPFPQSAGKGGPQEEVTRGPQGALGRLRELCRRWLRPEVHTKEQMLTMLPKEIQAWLQEHRPESSEEAAALVEDLTQTLQDSDFEIQSENGENCNQD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 54993 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title