missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93105-0.02ml
This item is not returnable.
View return policy
Descrizione
ZSCAN2 Polyclonal antibody specifically detects ZSCAN2 in Human samples. It is validated for Western Blot
Specifica
| ZSCAN2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ20595, zfp-29, ZFP29, zinc finger and SCAN domain containing 2, zinc finger and SCAN domain-containing protein 2, Zinc finger protein 29 homolog, Zinc finger protein 854, ZNF854zinc finger protein 29 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human ZSCAN2 (NP_870992.2). GPESEPFPQSAGKGGPQEEVTRGPQGALGRLRELCRRWLRPEVHTKEQMLTMLPKEIQAWLQEHRPESSEEAAALVEDLTQTLQDSDFEIQSENGENCNQD | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54993 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto