missing translation for 'onlineSavingsMsg'
Learn More

ZNF687 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

189.00€ - 546.00€

Specifications

Antigen ZNF687
Concentration 1 mg/mL
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18696005
View Documents
Novus Biologicals
NBP2-41128-0.025mg
0.025 mg
189.00€
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18000434
View Documents
Novus Biologicals
NBP2-41128
0.1 mg
546.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF687 Polyclonal specifically detects ZNF687 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ZNF687
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS with 0.02% Sodium Azide
57592
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZNF687. The immunogen is located within the first 50 amino acids of ZNF687. Amino Acid Squence: HSGPEENEGPGGPGKPE
Primary
ZNF687 antibody is human specific. At least three isoforms of ZNF687 are known to exist; this antibody will detect the the two largest isoforms.
1 mg/mL
Polyclonal
Rabbit
Human
KIAA1441DKFZp781I1719, zinc finger protein 687
ZNF687
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.