missing translation for 'onlineSavingsMsg'
Learn More

ZNF687 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-41128-0.025mg

 View more versions of this product

Product Code. 18696005

  • 189.00€ / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZNF687 Polyclonal specifically detects ZNF687 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF687
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
ZNF687
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZNF687. The immunogen is located within the first 50 amino acids of ZNF687. Amino Acid Squence: HSGPEENEGPGGPGKPE
0.025 mg
Primary
ZNF687 antibody is human specific. At least three isoforms of ZNF687 are known to exist; this antibody will detect the the two largest isoforms.
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
1 mg/mL
ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
KIAA1441DKFZp781I1719, zinc finger protein 687
Rabbit
Affinity Purified
RUO
57592
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.