missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | WRN |
|---|---|
| Concentration | 0.56 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16883845
|
Proteintech
25269-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16873845
|
Proteintech
25269-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the RecQ subfamily and the DEAH (Asp-Glu-Ala-His) subfamily of DNA and RNA helicases. DNA helicases are involved in many aspects of DNA metabolism, including transcription, replication, recombination, and repair. This protein contains a nuclear localization signal in the C-terminus and shows a predomit nucleolar localization. It possesses an intrinsic 3′ to 5′ DNA helicase activity, and is also a 3′ to 5′ exonuclease. Based on interactions between this protein and Ku70/80 heterodimer in DNA end processing, this protein may be involved in the repair of double strand DNA breaks. Defects in this gene are the cause of Werner syndrome, an autosomal recessive disorder characterized by premature aging.Specifications
| WRN | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q14191 | |
| 7486 | |
| Peptide | |
| Primary | |
| -20°C | |
| WRN |
| 0.56 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DNA helicase, RecQ like type 3, Exonuclease WRN, RecQ protein like 2, RECQ3, RECQL2, RECQL3, WRN | |
| WRN | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title