missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25269-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the RecQ subfamily and the DEAH (Asp-Glu-Ala-His) subfamily of DNA and RNA helicases. DNA helicases are involved in many aspects of DNA metabolism, including transcription, replication, recombination, and repair. This protein contains a nuclear localization signal in the C-terminus and shows a predomit nucleolar localization. It possesses an intrinsic 3′ to 5′ DNA helicase activity, and is also a 3′ to 5′ exonuclease. Based on interactions between this protein and Ku70/80 heterodimer in DNA end processing, this protein may be involved in the repair of double strand DNA breaks. Defects in this gene are the cause of Werner syndrome, an autosomal recessive disorder characterized by premature aging.Specifications
| WRN | |
| Polyclonal | |
| Unconjugated | |
| WRN | |
| DNA helicase, RecQ like type 3, Exonuclease WRN, RecQ protein like 2, RECQ3, RECQL2, RECQL3, WRN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7486 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.56 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14191 | |
| WRN | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction