missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF-1 Antibody (1G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51658-0.025ml
This item is not returnable.
View return policy
Description
WIF-1 Monoclonal specifically detects WIF-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| WIF-1 | |
| Monoclonal | |
| Unconjugated | |
| WIF-1, WNT inhibitory factor 1 | |
| Mouse | |
| 42 kDa | |
| 0.025 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 11197 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 1G5 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| WIF1 | |
| Purified recombinant fragment of human WIF1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction