missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF-1 Antibody (1G5), Novus Biologicals™
Mouse Monoclonal Antibody
217.00€
Specifications
| Antigen | WIF-1 |
|---|---|
| Clone | 1G5 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
WIF-1 Monoclonal specifically detects WIF-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| WIF-1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 11197 | |
| Purified recombinant fragment of human WIF1 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1G5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction, Wnt Signaling Pathway | |
| WIF-1, WNT inhibitory factor 1 | |
| WIF1 | |
| IgG1 | |
| Unpurified | |
| 42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title