Learn More
DWK Life Sciences Wheaton™ Clear Standard Wide-Mouth Bottles: With Polyvinyl-Lined Caps
Product Code. p-4908473 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
2000 mL
4000 mL
Unit Size:
Pack of 24
Pack of 4
Pack of 6
Product Code. 12359829
Brand: DWK Life Sciences W216924
Only null left
Add to basket
Add to basket
Ideal for the storage of dry materials and specimens
  • Clear for easy viewing of contents
  • Wide mouth for easy access to contents
  • Rounded shoulders
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear Type III soda-lime glass
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 oz., 125mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Clear
Diameter (English) 2.04 in.
Material Soda Lime Glass
Closure Color White
Height (Metric) 84 mm
Height (English) 3.3 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 48-400 mm
Diameter (Metric) 52 mm
Closure Type Screw Cap