Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps
Product Code. 12319749 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Packaging:
Convenience pack
Valu-bulk pack
Unit Size:
Pack of 108
Pack of 12
Pack of 160
Pack of 24
Pack of 288
Pack of 48
Product Code. 12319749
Brand: DWK Life Sciences W216826
Only null left
Add to basket
Add to basket
Ideal for solvent, chemical and sample storage
  • Narrow mouth enables liquids to be carefully poured to prevent spillage
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
  • Available in partitioned Convenience or Valu-Bulk™ cartons
Order Info
Caps can be ordered separately.
Compliance
USP Type-III soda-lime glass
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Clear
Diameter (English) 1.88 in.
Material Soda Lime Glass
Packaging Valu-bulk pack
Closure Color White
Certifications/Compliance EPA, FDA
Height (Metric) 112 mm
Height (English) 4.4 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 22-400 mm
Diameter (Metric) 48 mm
Closure Type Screw Cap