Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds with Polyvinyl-lined Caps
Product Code. 10225262 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Packaging:
Convenience pack
Valu-bulk Pk.
Valu-bulk pack
Unit Size:
Pack of 12
Pack of 160
Pack of 24
Pack of 288
Pack of 30
Pack of 48
Pack of 60
Product Code. 10225262
Brand: DWK Life Sciences W216840
Amber Type III soda lime glass protects light-sensitive solutions
  • Available in partitioned Convenience Pack or Valu-Bulk™ cartons
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Each case of Convenience Pack bottles includes Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
USP Type III soda-lime glass
TRUSTED_SUSTAINABILITY
Specifications
Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Amber
Diameter (English) 2.95 in.
Material Soda Lime Glass
Packaging Convenience pack
Closure Color White
Height (Metric) 168 mm
Height (English) 6.6 in.
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 28-400 mm
Diameter (Metric) 75 mm
Closure Type Screw Cap