missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP2 Mouse anti-Human, CoraLite 594, Clone: 2B4G7, Proteintech
Mouse Monoclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | WBP2 |
|---|---|
| Clone | 2B4G7 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17771419
|
Proteintech
CL594-66585-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17619966
|
Proteintech
CL594-66585-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WBP2 Monoclonal antibody apecifically detetects WBP2 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| WBP2 | |
| Immunofluorescence | |
| CoraLite 594 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 23558 | |
| WBP2 fusion protein Ag2661 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 2B4G7 | |
| Monoclonal | |
| Liquid | |
| Human | |
| BC010616 | |
| WBP2 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title