missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP2 Mouse anti-Human, CoraLite 594, Clone: 2B4G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66585-20UL
This item is not returnable.
View return policy
Description
WBP2 Monoclonal antibody apecifically detetects WBP2 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| WBP2 | |
| Monoclonal | |
| 1000 ug/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| WBP2 | |
| WBP2 fusion protein Ag2661 | |
| 20 μL | |
| 23558 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Immunofluorescence | |
| 2B4G7 | |
| CoraLite 594 | |
| BC010616 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction