missing translation for 'onlineSavingsMsg'
Learn More

Viral HIV-1 Gag p24 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

190.05€ - 423.15€

Specifications

Antigen HIV-1 Gag p24
Clone 1042904
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18710857
GREENER_CHOICE
View Documents
R&D Systems
MAB73603-SP
25 μg
ONLINE EXCLUSIVE PRICE
201.00€
190.05€ / 25µg
Save 10.95€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18792917
GREENER_CHOICE
View Documents
R&D Systems
MAB73603-100
100 μg
ONLINE EXCLUSIVE PRICE
448.00€
423.15€ / 100µg
Save 24.85€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot
Specifications

Specifications

HIV-1 Gag p24
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
155030
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1042904
Western Blot
Unconjugated
Mouse
Virus
Q77YG1
E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.