Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB73603-100
 View more versions of this product
Product Code. 18792917
  • ONLINE EXCLUSIVE PRICE
    448.00€
    423.15€ / 100µg
    Save 24.85€
    5% Off
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HIV-1 Gag p24
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
155030
Virus
Purified
Western Blot
1042904
Western Blot 1 ug/mL
Q77YG1
E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.