missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Antibody (BV14) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 6 publications
208.00€ - 595.00€
Specifications
| Antigen | VE-Cadherin |
|---|---|
| Clone | BV14 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006514
|
Novus Biologicals
NBP1-43347-0.025MG |
0.025 mg |
208.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18062424
|
Novus Biologicals
NBP1-43347-0.5MG |
0.5 mg |
595.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| VE-Cadherin | |
| 0.5 mg/mL | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Mouse | |
| P55284 | |
| 1003 | |
| The immunogen for this antibody was CD144. | |
| Primary | |
| Store at 4C. Do not freeze. |
| BV14 | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Rat | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| CDH5 | |
| IgG2b | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title