missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Antibody (BV14) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-43347-0.5MG
This item is not returnable.
View return policy
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| VE-Cadherin | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P55284 | |
| CDH5 | |
| The immunogen for this antibody was CD144. | |
| 0.5 mg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| 1003 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| BV14 | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction