missing translation for 'onlineSavingsMsg'
Learn More

VE-Cadherin Antibody (BV14) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 6 publications

Brand:  Novus Biologicals NBP1-43347-0.025MG

163.20 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 18006514

  • 208.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VE-Cadherin
Monoclonal
0.5 mg/mL
Western Blot 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P55284
CDH5
The immunogen for this antibody was CD144.
0.025 mg
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
1003
Store at 4C. Do not freeze.
Western Blot, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry
BV14
Unconjugated
PBS, pH 7.2 with 0.09% Sodium Azide
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin
Rat
Affinity Purified
RUO
Primary
Mouse
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only