missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP3/Cellubrevin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
204.00€
Specifications
| Antigen | VAMP3/Cellubrevin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
VAMP3/Cellubrevin Polyclonal specifically detects VAMP3/Cellubrevin in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| VAMP3/Cellubrevin | |
| Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 | |
| Polyclonal | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
| VAMP3 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P63024 | |
| 9341 | |
| Sythetic peptide corresponding to residues M(1) S T G V P S G S S A A T G S N R R(18) C of mouse VAMP-3. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title