Learn More
VAMP3/Cellubrevin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NB300-510-0.025mg
 View more versions of this product
Product Code. 18526764
  • 204.00€ / 25µg
Description
VAMP3/Cellubrevin Polyclonal specifically detects VAMP3/Cellubrevin in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VAMP3/Cellubrevin
Polyclonal
Unconjugated
PBS with 1mg/ml BSA with 0.05% Sodium Azide
CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin)
Rabbit
Affinity Purified
RUO
Primary
Human
IgG
Western Blot
1 mg/mL
Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100
P63024
VAMP3
Sythetic peptide corresponding to residues M(1) S T G V P S G S S A A T G S N R R(18) C of mouse VAMP-3.
0.025 mg
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission
9341
Store at -20°C. Avoid freeze/thaw cycles.