missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-98492
This item is not returnable.
View return policy
Description
STRA8 Polyclonal specifically detects STRA8 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.
Specifications
| STRA8 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| stimulated by retinoic acid gene 8 homolog (mouse), stimulated by retinoic acid gene 8 protein homolog | |
| Rabbit | |
| 37 kDa | |
| 100 μL | |
| Primary | |
| Canine: 77%; Porcine: 77%. | |
| Human, Mouse, Canine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q7Z7C7 | |
| STRA8 | |
| The immunogen for this antibody is STRA8 - C-terminal region (NP_872295). Peptide sequence PEEKFQLYMQIINFFKGLSCANTQVKQEASFPVDEEMIMLQCTETFDDED. | |
| Affinity purified | |
| RUO | |
| 346673 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction