missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V alpha 24 J alpha 18 TCR Antibody (6B11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NBP2-00267
This item is not returnable.
View return policy
Description
V alpha 24 J alpha 18 TCR Monoclonal specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Cell Surface), CyTOF-ready.
Specifications
| V alpha 24 J alpha 18 TCR | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16867051), Immunohistochemistry-Frozen reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready | |
| iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell | |
| Peptide epitope in the CDR3 region of V alpha 24 | |
| 0.1 mg | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 6B11 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction