missing translation for 'onlineSavingsMsg'
Learn More

V alpha 24 J alpha 18 TCR Antibody (6B11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 12 publications

237.00€ - 473.00€

Specifications

Antigen V alpha 24 J alpha 18 TCR
Clone 6B11
Concentration 1.0 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16867051), Immunohistochemistry-Frozen reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18005286
View Documents
Novus Biologicals
NBP2-00267SS
0.025 mg
237.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18052177
View Documents
Novus Biologicals
NBP2-00267
0.1 mg
473.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

V alpha 24 J alpha 18 TCR Monoclonal specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Cell Surface), CyTOF-ready.
Specifications

Specifications

V alpha 24 J alpha 18 TCR
1.0 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Human
iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell
IgG1 κ
Protein G purified
6B11
Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16867051), Immunohistochemistry-Frozen reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready
Monoclonal
Purified
RUO
PBS with 0.02% Sodium Azide
Peptide epitope in the CDR3 region of V alpha 24
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.