missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00€ - 322.00€
Specifications
| Antigen | UQCRH |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687527
|
Novus Biologicals
NBP3-12568-50ul |
50 μg |
183.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688696
|
Novus Biologicals
NBP3-12568-100ul |
100 μg |
322.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UQCRH Polyclonal antibody specifically detects UQCRH in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| UQCRH | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH7.0 | |
| 7388 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Complex III subunit 6, Complex III subunit VIII, cytochrome b-c1 complex subunit 6, mitochondrial, Cytochrome c1 non-heme 11 kDa protein, MGC111572, Mitochondrial hinge protein, QCR6, Ubiquinol-cytochrome c reductase complex 11 kDa protein, ubiquinol-cytochrome c reductase hinge protein, ubiquinol-cytochrome c reductase, complex III subunit VIII, UQCR8 | |
| Produced in rabbits immunized with E. coli-derived Human UQCRH fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title