missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12568-50ul
This item is not returnable.
View return policy
Description
UQCRH Polyclonal antibody specifically detects UQCRH in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UQCRH | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Complex III subunit 6, Complex III subunit VIII, cytochrome b-c1 complex subunit 6, mitochondrial, Cytochrome c1 non-heme 11 kDa protein, MGC111572, Mitochondrial hinge protein, QCR6, Ubiquinol-cytochrome c reductase complex 11 kDa protein, ubiquinol-cytochrome c reductase hinge protein, ubiquinol-cytochrome c reductase, complex III subunit VIII, UQCR8 | |
| Produced in rabbits immunized with E. coli-derived Human UQCRH fragment | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7388 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction