Learn More
UOP Method 744 Standard-8 Components, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| n-Heptane | 142-82-5 | 25% (w/w) |
| Toluene | 108-88-3 | 20% (w/w) |
| Benzene | 71-43-2 | 15% (w/w) |
| 1,2,3,4-Tetramethylbenzene | 488-23-3 | 10% (w/w) |
| 2-Ethyltoluene | 611-14-3 | 10% (w/w) |
| m-Xylene | 108-38-3 | 6.7% (w/w) |
| p-Xylene | 106-42-3 | 6.7% (w/w) |
| o-Xylene | 95-47-6 | 6.6% (w/w) |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | UOP Method 744 Standard |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | UOP Method 744 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solvent or Matrix | Neat |
| Quantity | 1 mL |
Product intended for laboratory use only