missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin B Antibody (rUBB/9476), Novus Biologicals™
Mouse Monoclonal Antibody
256.00€ - 484.00€
Specifications
| Antigen | Ubiquitin B |
|---|---|
| Clone | rUBB/9476 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30290830
|
Novus Biologicals
NBP3-44622-20ug |
20 μg |
256.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30304587
|
Novus Biologicals
NBP3-44622-100ug |
100 μg |
484.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ubiquitin B Monoclonal antibody specifically detects Ubiquitin B in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| Ubiquitin B | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
| Recombinant fragment of human Ubiquitin protein (around aa 1-119) (Uniprot: P0G47; P0G48; P62979; P62987; P62988). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| rUBB/9476 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Apoptosis, Cell Biology, Neurodegeneration, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 7314 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title