missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin B Antibody (rUBB/9476), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44622-100ug
This item is not returnable.
View return policy
Description
Ubiquitin B Monoclonal antibody specifically detects Ubiquitin B in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Ubiquitin B | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| rUBB/9476 | |
| Unconjugated | |
| FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
| Recombinant fragment of human Ubiquitin protein (around aa 1-119) (Uniprot: P0G47; P0G48; P62979; P62987; P62988). Exact sequence is proprietary. | |
| 100 μg | |
| Alzheimers Research, Apoptosis, Cell Biology, Neurodegeneration, Neuroscience | |
| 7314 | |
| Store at 4⪚C. | |
| IgG2b κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur