missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYW3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€
Specifications
| Antigen | TYW3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TYW3 Polyclonal specifically detects TYW3 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
| TYW3 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q6IPR3 | |
| 127253 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human TYW3. The immunogen is located within amino acids 160 - 210 of TYW3. Amino Acid Squence: ANQKMEENKKRIERFYN | |
| Primary | |
| Store at 4°C. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| C1orf171, chromosome 1 open reading frame 171, EC 2.1.1, EC 2.1.1.-, FLJ40918, tRNA wybutosine-synthesizing protein 3 homolog, tRNA-yW synthesizing protein 3 homolog (S. cerevisiae), tRNA-yW-synthesizing protein 3 | |
| TYW3 | |
| IgG | |
| Affinity Purified | |
| TYW3 antibody is predicted to not cross-react with other TYW protein family members. At least two isoforms of TYW3 are known to exist; this antibody will detect both isoforms. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title