missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYW3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76584-0.025mg
This item is not returnable.
View return policy
Description
TYW3 Polyclonal specifically detects TYW3 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| TYW3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C1orf171, chromosome 1 open reading frame 171, EC 2.1.1, EC 2.1.1.-, FLJ40918, tRNA wybutosine-synthesizing protein 3 homolog, tRNA-yW synthesizing protein 3 homolog (S. cerevisiae), tRNA-yW-synthesizing protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 127253 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000 | |
| Q6IPR3 | |
| TYW3 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human TYW3. The immunogen is located within amino acids 160 - 210 of TYW3. Amino Acid Squence: ANQKMEENKKRIERFYN | |
| 0.025 mg | |
| Primary | |
| TYW3 antibody is predicted to not cross-react with other TYW protein family members. At least two isoforms of TYW3 are known to exist; this antibody will detect both isoforms. | |
| Store at 4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction