missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
189.00€
Specifications
| Antigen | TWEAK/TNFSF12 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5-10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TWEAK/TNFSF12 Polyclonal specifically detects TWEAK/TNFSF12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| TWEAK/TNFSF12 | |
| Western Blot 0.5-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5-10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| O43508 | |
| 8742 | |
| Antibody was raised against recombinant human TWEAK protein. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| TNFSF12 | |
| IgG | |
| Protein A purified | |
| 32 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title