missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-76695-0.025mg
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Polyclonal specifically detects TWEAK/TNFSF12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| TWEAK/TNFSF12 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| Rabbit | |
| 32 kDa | |
| 0.025 mg | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| 8742 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5-10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| O43508 | |
| TNFSF12 | |
| Antibody was raised against recombinant human TWEAK protein. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction