missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trk (pan) Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 472.00€
Specifications
| Antigen | Trk (pan) |
|---|---|
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698727
|
Novus Biologicals
NBP3-12168 |
0.5 mL |
472.00€
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600268
|
Novus Biologicals
NBP3-12168-0.1ml |
0.1 mL |
190.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Trk (pan) Polyclonal antibody specifically detects Trk (pan) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Trk (pan) | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 | |
| 4914 | |
| Primary | |
| Store at 4°C. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| High affinity nerve growth factor receptor;MTC;NTRK1;Nucleoprotein TPR;p140-TrkA;Protein TFG;TRK;TRK1;TRKA;Trk-A;Tyrosine-protein kinase receptor | |
| Peptide-KLH conjugate with human Trk A (777-796aa) | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title