missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trk (pan) Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12168-0.1ml
This item is not returnable.
View return policy
Description
Trk (pan) Polyclonal antibody specifically detects Trk (pan) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Trk (pan) | |
| Polyclonal | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4914 | |
| Store at 4°C. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| High affinity nerve growth factor receptor;MTC;NTRK1;Nucleoprotein TPR;p140-TrkA;Protein TFG;TRK;TRK1;TRKA;Trk-A;Tyrosine-protein kinase receptor | |
| Peptide-KLH conjugate with human Trk A (777-796aa) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction