missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM21 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | TRIM21 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18097305
|
Proteintech
CL488-12108-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18077305
|
Proteintech
CL488-12108-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TRIM21 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM21. This antibody recognizes human antigen. The TRIM21 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| TRIM21 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| 52 kDa Ro protein, RING finger protein 81, RNF81, Ro(SS A), RO52, SS A, SSA, SSA1, TRIM21, tripartite motif containing 21 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 6737 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title