Learn More
TRIM21 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-12108-20UL
 View more versions of this product
Product Code. 18097305
  • 145.00€ / 20µL
Description
The TRIM21 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM21. This antibody recognizes human antigen. The TRIM21 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
TRIM21
1000 μg/mL
52 kDa Ro protein, RING finger protein 81, RNF81, Ro(SS A), RO52, SS A, SSA, SSA1, TRIM21, tripartite motif containing 21
20 μL
6737
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa