Learn More
TRIM21 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-12108-20UL
Description
The TRIM21 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRIM21. This antibody recognizes human antigen. The TRIM21 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| TRIM21 | |
| 1000 μg/mL | |
| 52 kDa Ro protein, RING finger protein 81, RNF81, Ro(SS A), RO52, SS A, SSA, SSA1, TRIM21, tripartite motif containing 21 | |
| 20 μL | |
| 6737 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |