missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAX Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 470.00€
Specifications
| Antigen | TRAX |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635880
|
Novus Biologicals
NBP2-93964-0.02ml |
0.02 mL |
188.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644641
|
Novus Biologicals
NBP2-93964-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAX Polyclonal antibody specifically detects TRAX in Human, Mouse samples. It is validated for Western BlotSpecifications
| TRAX | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Direct Reversal of DNA Damage | |
| PBS (pH 7.3), 50% glycerol | |
| 7257 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DISC1, translin-associated factor Xdisrupted in schizophrenia 1, translin-like protein, TRAXtranslin-associated protein X | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human TSNAX (NP_005990.1). MSNKEGSGGFRKRKHDNFPHNQRREGKDVNSSSPVMLAFKSFQQELDARHDKYERLVKLSRDITVESKRTIFLLHRITSAPDMEDILTESEIKLDGVRQKIFQVAQELSGEDMHQFHRAITTGLQEYVEAVSFQHFIKTRSLISMDEINKQLIFTTEDNGKENKTPSSDAQDKQFGTWRLRVTPVDYLLGVADLTGELMRMCINSVGNGDIDTPFEVSQFLRQVYDGFSFIGNTGPYEVSKKLYTLKQSLAKVENACYALKVRGSEIPKHMLADVFSVKTEMIDQEEGIS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title