missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAX Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93964-0.02ml
This item is not returnable.
View return policy
Description
TRAX Polyclonal antibody specifically detects TRAX in Human, Mouse samples. It is validated for Western Blot
Specifications
| TRAX | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DISC1, translin-associated factor Xdisrupted in schizophrenia 1, translin-like protein, TRAXtranslin-associated protein X | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human TSNAX (NP_005990.1). MSNKEGSGGFRKRKHDNFPHNQRREGKDVNSSSPVMLAFKSFQQELDARHDKYERLVKLSRDITVESKRTIFLLHRITSAPDMEDILTESEIKLDGVRQKIFQVAQELSGEDMHQFHRAITTGLQEYVEAVSFQHFIKTRSLISMDEINKQLIFTTEDNGKENKTPSSDAQDKQFGTWRLRVTPVDYLLGVADLTGELMRMCINSVGNGDIDTPFEVSQFLRQVYDGFSFIGNTGPYEVSKKLYTLKQSLAKVENACYALKVRGSEIPKHMLADVFSVKTEMIDQEEGIS | |
| 0.02 mL | |
| Direct Reversal of DNA Damage | |
| 7257 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction